Assay Details
| Gene Symbol: Ndfip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022996.1 | NP_075372.1 | 5 - 6 | 633 | 84 | |
| GenBank mRNA | AF220209.1 | - | 5 - 6 | 671 | 84 | |
| AK050560.1 | - | 5 - 6 | 640 | 84 | ||
| BC020359.1 | - | 5 - 6 | 620 | 84 | ||
| BC026372.1 | - | 5 - 6 | 633 | 4162070 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndfip1
Genomic Map