Assay Details
| Gene Symbol: Tars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163617.1 | NP_001157089.1 | 10 - 11 | 1395 | 79 | |
| NM_001163618.1 | NP_001157090.1 | 8 - 9 | 1152 | 79 | ||
| NM_027931.3 | NP_082207.2 | 10 - 11 | 1395 | 79 | ||
| GenBank mRNA | AK011540.1 | - | 10 - 11 | 1281 | 79 | |
| AK080690.1 | - | 10 - 11 | 80 | |||
| AK142685.1 | - | 10 - 11 | 1290 | 79 | ||
| BC008995.1 | - | 10 - 11 | 1280 | 3157108 | 79 | |
| BC067392.1 | - | 4 - 5 | 2503 | 79 | ||
| BC147501.1 | - | 10 - 11 | 1279 | 9056383 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tars2
Genomic Map