Assay Details
| Gene Symbol: Ttc39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145948.1 | NP_001139420.1 | 13 - 14 | 1205 | 85 | |
| NM_153392.2 | NP_700441.2 | 13 - 14 | 1214 | 85 | ||
| GenBank mRNA | AK048659.1 | - | 13 - 14 | 1215 | 85 | |
| AK076641.1 | - | 13 - 14 | 1167 | 85 | ||
| AK172951.1 | - | 13 - 14 | 1137 | 85 | ||
| BC023883.1 | - | 13 - 14 | 1213 | 5150841 | 85 | |
| BC026660.1 | - | 13 - 14 | 1155 | 4988900 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc39a
Genomic Map