Assay Details
| Gene Symbol: Hmgxb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134134.2 | NP_598895.2 | 11 - 12 | 2204 | 76 | |
| NM_178277.1 | NP_840061.1 | 10 - 11 | 2299 | 76 | ||
| GenBank mRNA | AK041866.1 | - | 11 - 12 | 2182 | 76 | |
| AK129085.1 | - | 7 - 8 | 1207 | 76 | ||
| AK132529.1 | - | 4 - 5 | 683 | 76 | ||
| AK144993.1 | - | 10 - 11 | 2052 | 76 | ||
| BC058089.1 | - | 8 - 9 | 1597 | 76 | ||
| BC060046.1 | - | 4 - 5 | 629 | 76 | ||
| BC066029.1 | - | 5 - 6 | 1435 | 6417061 | 76 | |
| BC079580.1 | - | 10 - 11 | 2247 | 5702202 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgxb3
Genomic Map