Assay Details
| Gene Symbol: Slc27a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081072.1 | NP_001074541.1 | 8 - 9 | 1770 | 70 | |
| GenBank mRNA | AK028699.1 | - | 8 - 9 | 1772 | 70 | |
| BC115435.1 | - | 8 - 9 | 1753 | 40094587 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a6
Genomic Map