Assay Details
| Gene Symbol: Alpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037294.1 | NP_001032371.1 | 9 - 10 | 6136 | 73 | |
| GenBank mRNA | AK132555.1 | - | 6 - 7 | 4464 | 73 | |
| AY044451.1 | - | 5 - 6 | 3858 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alpk2
Genomic Map