Assay Details
| Gene Symbol: Alpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037294.1 | NP_001032371.1 | 11 - 12 | 6435 | 91 | |
| GenBank mRNA | AK132555.1 | - | 8 - 9 | 4763 | 91 | |
| AY044451.1 | - | 7 - 8 | 4157 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alpk2
Genomic Map