Assay Details
| Gene Symbol: Pcyox1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172832.4 | NP_766420.1 | 6 | 1628 | 90 | |
| GenBank mRNA | BC060677.1 | - | 6 | 1618 | 6404883 | 90 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31759276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31759279 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31759277 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248387 | AK050006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P248387 | AK050006.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160756 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142874 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94210 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119001 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178914 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248387 | AK050006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P248387 | AK050006.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31759272 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178914 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119001 | - | - |
| Whole Mouse Genome G4122A | A_51_P248387 | AK050006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248387 | AK050006.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127755 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119001 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178914 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13685523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13685522 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13685530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13685525 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31759279 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31759277 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31759276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31759160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13923189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14120427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13685532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14027745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14135440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13945356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13625786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14135439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13685534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13859451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31759287 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10459233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459234 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455380_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10459229 | - | - |
| Mouse Genome 430 2.0 Array | 1455380_at | - | - |
| Mouse Genome 430 2.0 Array | 1455380_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240334_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240334_PM_at | - | - |
| Mouse Expression Set 430 | 1455380_at | - | - |
| Mouse Expression Set 430 | 1455380_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455380_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455380_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 331057 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4810692 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810692 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684577 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684577 | - | - |
| mouse-6 v1.1 (Illumina) | scl51357.6.1_32-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810692 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51357.6.1_32-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810692 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2684575 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2684575 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl51357.6.1_32-S | - | - |

Pcyox1l
Genomic Map