Assay Details
| Gene Symbol: Mxra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB052097.1 | - | 4 - 5 | 511 | 62 | |
| EU849596.1 | - | 4 - 5 | 471 | 62 | ||
| EU849597.1 | - | 4 - 5 | 462 | 62 | ||
More Information
Set Membership: |
|

Mxra8
Genomic Map