Assay Details
| Gene Symbol: Ankzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026187.5 | NP_080463.3 | 6 - 7 | 1185 | 72 | |
| NR_051998.1 | 6 - 7 | 1216 | 72 | |||
| GenBank mRNA | AK004948.2 | - | 4 - 5 | 1615 | 72 | |
| AK156255.1 | - | 6 - 7 | 1149 | 72 | ||
| AK157405.1 | - | 6 - 7 | 1149 | 72 | ||
| BC071240.1 | - | 6 - 7 | 1115 | 72 | ||
| BC150879.1 | - | 6 - 7 | 1135 | 9087790 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156255.1 transcripts.
Set Membership: |
|

Ankzf1
Genomic Map