Assay Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008985.2 | NP_033011.2 | 6 - 7 | 1074 | 99 | |
| GenBank mRNA | AF332083.1 | - | 3 - 4 | 678 | 99 | |
| AF332084.1 | - | 3 - 4 | 678 | 99 | ||
| AK041296.1 | - | 6 - 7 | 1080 | 99 | ||
| AK076223.1 | - | 6 - 7 | 1055 | 99 | ||
| AK163141.1 | - | 6 - 7 | 1052 | 99 | ||
| AK220467.1 | - | 6 - 7 | 1032 | 99 | ||
| BC068165.1 | - | 6 - 7 | 1006 | 99 | ||
| DQ832283.1 | - | 6 - 7 | 1051 | 99 | ||
| U11812.1 | - | 6 - 7 | 992 | 99 | ||
| X74438.1 | - | 6 - 7 | 1091 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptprn
Genomic Map