Assay Details
| Gene Symbol: Bcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024468.3 | NP_001019639.1 | 6 - 7 | 914 | 69 | |
| NM_007532.5 | NP_031558.3 | 6 - 7 | 933 | 69 | ||
| GenBank mRNA | AK012821.1 | - | 6 - 7 | 708 | 69 | |
| AK013888.1 | - | 6 - 7 | 870 | 69 | ||
| AK036309.1 | - | 6 - 7 | 915 | 69 | ||
| AK138468.1 | - | 6 - 7 | 827 | 69 | ||
| AK166971.1 | - | 6 - 7 | 869 | 69 | ||
| AK167232.1 | - | 6 - 7 | 860 | 69 | ||
| AK167368.1 | - | 6 - 7 | 779 | 69 | ||
| AK168249.1 | - | 6 - 7 | 963 | 69 | ||
| BC053706.1 | - | 6 - 7 | 865 | 30063465 | 69 | |
| U42443.1 | - | 5 - 6 | 670 | 69 | ||
| X17502.1 | - | 6 - 7 | 857 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcat1
Genomic Map