Assay Details
| Gene Symbol: Gpr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020337.1 | - | 1 - 2 | 76 | 115 | |
| AK035091.1 | - | 1 - 2 | 336 | 115 | ||
| AK054151.1 | - | 1 - 2 | 363 | 115 | ||
More Information
Set Membership: |
|

Gpr45
Genomic Map