Assay Details
| Gene Symbol: Atg4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145967.1 | NP_001139439.1 | 3 - 4 | 363 | 87 | |
| NM_175029.3 | NP_778194.3 | 3 - 4 | 745 | 87 | ||
| GenBank mRNA | AK134838.1 | - | 3 - 4 | 512 | 87 | |
| AK156463.1 | - | 3 - 4 | 515 | 87 | ||
| AK170747.1 | - | 3 - 4 | 502 | 87 | ||
| BB851037.1 | - | 3 - 4 | 364 | 87 | ||
| BC058981.1 | - | 3 - 4 | 745 | 6827140 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atg4c
Genomic Map