Assay Details
| Gene Symbol: Icmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133788.2 | NP_598549.1 | 3 - 4 | 616 | 66 | |
| NR_037908.1 | 4 - 5 | 725 | 66 | |||
| GenBank mRNA | AF209926.1 | - | 3 - 4 | 398 | 66 | |
| AK154135.1 | - | 3 - 4 | 548 | 66 | ||
| AK154410.1 | - | 4 - 5 | 639 | 66 | ||
| BC057132.1 | - | 4 - 5 | 635 | 66 | ||
| BC110695.1 | - | 3 - 4 | 603 | 6842587 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Icmt
Genomic Map