Assay Details
| Gene Symbol: LOC102637577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047309.1 | - | 1 - 2 | 68 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear factor I/A | Nfia |
1110047K16Rik, 9430022M17Rik, CTF, NF1-A, NF1A |
Anti-sense | Chr. 4 - 97581717 - 98118876 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 0610025J13 gene | 0610025J13Rik | B430316H17Rik | 563.3kb | Anti-sense | Chr. 4 - 98320611 - 98331960 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102637577
Genomic Map