Assay Details
| Gene Symbol: Alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102446.1 | NP_001095916.1 | 9 - 10 | 1477 | 80 | |
| NM_009653.3 | NP_033783.1 | 9 - 10 | 1522 | 80 | ||
| GenBank mRNA | AK002642.1 | - | 9 - 10 | 1493 | 80 | |
| AK077610.1 | - | 9 - 10 | 1494 | 80 | ||
| AK165257.1 | - | 9 - 10 | 1485 | 80 | ||
| AK165262.1 | - | 9 - 10 | 1497 | 80 | ||
| AK172094.1 | - | 9 - 10 | 1486 | 80 | ||
| BC024575.1 | - | 5 - 6 | 890 | 80 | ||
| BC150870.1 | - | 9 - 10 | 1469 | 9087781 | 80 | |
| M15268.1 | - | 9 - 10 | 1435 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alas2
Genomic Map