Assay Details
| Gene Symbol: Impact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008378.2 | NP_032404.1 | 8 - 9 | 747 | 60 | |
| GenBank mRNA | AK138136.1 | - | 8 - 9 | 718 | 60 | |
| BC020524.1 | - | 8 - 9 | 723 | 4223620 | 60 | |
| D87973.1 | - | 8 - 9 | 747 | 60 | ||
More Information
Set Membership: |
|

Impact
Genomic Map