Assay Details
| Gene Symbol: Sfxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053198.3 | NP_444428.3 | 11 - 12 | 744 | 67 | |
| GenBank mRNA | AF325263.1 | - | 11 - 12 | 686 | 67 | |
| AK148530.1 | - | 10 - 11 | 713 | 67 | ||
| BC058211.1 | - | 12 - 13 | 936 | 67 | ||
| BC089535.1 | - | 11 - 12 | 726 | 6589805 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sfxn4
Genomic Map