Assay Details
| Gene Symbol: Etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035231.1 | - | 2 - 3 | 270 | 75 | |
| AK078773.1 | - | 3 - 4 | 460 | 75 | ||
| AK154569.1 | - | 3 - 4 | 548 | 75 | ||
| AK155150.1 | - | 3 - 4 | 733 | 75 | ||
| AK155752.1 | - | 2 - 3 | 282 | 75 | ||
| AK157265.1 | - | 3 - 4 | 402 | 75 | ||
| BC052163.1 | - | 3 - 4 | 421 | 6513848 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078773.1, AK154569.1, AK155150.1, AK157265.1, BC052163.1 transcripts.
Set Membership: |
|

Etv6
Genomic Map