Assay Details
| Gene Symbol: Etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035231.1 | - | 5 - 6 | 1095 | 83 | |
| AK078773.1 | - | 6 - 7 | 1288 | 83 | ||
| AK154569.1 | - | 6 - 7 | 1376 | 83 | ||
| AK155150.1 | - | 6 - 7 | 1561 | 83 | ||
| AK155752.1 | - | 5 - 6 | 1111 | 83 | ||
| AK157265.1 | - | 6 - 7 | 1230 | 83 | ||
| BC052163.1 | - | 6 - 7 | 1249 | 6513848 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etv6
Genomic Map