Assay Details
| Gene Symbol: Arhgdib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK169733.1 | - | 2 - 3 | 257 | 70 | |
| BC031763.1 | - | 2 - 3 | 212 | 4923219 | 70 | |
| L07918.1 | - | 2 - 3 | 213 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgdib
Genomic Map