Assay Details
| Gene Symbol: Arhgdib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK169733.1 | - | 3 - 4 | 341 | 81 | |
| BC031763.1 | - | 3 - 4 | 296 | 4923219 | 81 | |
| L07918.1 | - | 3 - 4 | 297 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgdib
Genomic Map