Assay Details
| Gene Symbol: Mrpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030116.2 | NP_084392.1 | 1 - 2 | 162 | 55 | |
| GenBank mRNA | AB049637.1 | - | 1 - 2 | 122 | 55 | |
| AK021196.1 | - | 1 - 2 | 181 | 55 | ||
| AK040055.1 | - | 1 - 2 | 163 | 55 | ||
| AK046099.1 | - | 1 - 2 | 162 | 55 | ||
| AK146237.1 | - | 163 | 55 | |||
| AK161563.1 | - | 1 - 2 | 166 | 55 | ||
| AK162023.1 | - | 1 - 2 | 162 | 55 | ||
| BC116385.1 | - | 1 - 2 | 162 | 40101776 | 55 | |
| BC116386.1 | - | 1 - 2 | 162 | 40101778 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl9
Genomic Map