| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600025969.mm.1 |
NM_001167696.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025969.mm.1 |
TC0600003311.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NM_001167694.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NM_001167695.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NM_001167696.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NM_001167697.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NM_001167699.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NM_001167700.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NM_008157.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NR_072990.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
NR_074084.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025960.mm.1 |
TC0600003311.mm.1 |
MTA TC |