Assay Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198967.5 | NP_945318.2 | 18 - 19 | 2762 | 78 | |
| GenBank mRNA | AK046396.1 | - | 17 - 18 | 2649 | 78 | |
| AK137545.1 | - | 18 - 19 | 2734 | 78 | ||
| BC023818.1 | - | 7 - 8 | 923 | 78 | ||
| BC145513.1 | - | 16 - 17 | 2094 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmtc1
Genomic Map