Assay Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198967.5 | NP_945318.2 | 14 - 15 | 2224 | 78 | |
| GenBank mRNA | AK046396.1 | - | 13 - 14 | 2111 | 78 | |
| AK137545.1 | - | 14 - 15 | 2196 | 78 | ||
| AK162239.1 | - | 15 - 16 | 1964 | 78 | ||
| BC023818.1 | - | 3 - 4 | 385 | 78 | ||
| BC145513.1 | - | 12 - 13 | 1556 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmtc1
Genomic Map