Assay Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198967.5 | NP_945318.2 | 16 - 17 | 2501 | 92 | |
| GenBank mRNA | AK046396.1 | - | 15 - 16 | 2388 | 92 | |
| AK137545.1 | - | 16 - 17 | 2473 | 92 | ||
| BC023818.1 | - | 5 - 6 | 662 | 92 | ||
| BC145513.1 | - | 14 - 15 | 1833 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmtc1
Genomic Map