Assay Details
| Gene Symbol: Ankzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267620.1 | NP_001254549.1 | 3 - 4 | 724 | 83 | |
| NM_026187.5 | NP_080463.3 | 3 - 4 | 724 | 83 | ||
| NR_051998.1 | 3 - 4 | 724 | 83 | |||
| NR_051999.1 | 2 - 3 | 617 | 83 | |||
| GenBank mRNA | AK084573.1 | - | 2 - 3 | 579 | 83 | |
| AK156255.1 | - | 3 - 4 | 688 | 83 | ||
| AK157405.1 | - | 3 - 4 | 688 | 83 | ||
| BC039910.1 | - | 3 - 4 | 687 | 83 | ||
| BC051449.1 | - | 3 - 4 | 704 | 83 | ||
| BC150879.1 | - | 3 - 4 | 674 | 9087790 | 83 | |
| BU054541.1 | - | 2 - 3 | 125 | 83 | ||
| DV062487.1 | - | 3 - 4 | 748 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156255.1 transcripts.
Set Membership: |
|

Ankzf1
Genomic Map