Assay Details
| Gene Symbol: Cfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007688.2 | NP_031714.1 | 3 - 4 | 517 | 123 | |
| GenBank mRNA | AK004659.1 | - | 3 - 4 | 510 | 123 | |
| AK147174.1 | - | 3 - 4 | 431 | 123 | ||
| BC007138.1 | - | 3 - 4 | 460 | 3590849 | 123 | |
| L29468.1 | - | 3 - 4 | 517 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449508 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449509 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449511 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449513 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100159 | L29468.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100159 | BC007138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100159 | AK147174.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100159 | NM_007688.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100159 | AK004659.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | L29468.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | AK162912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | BC007138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | NM_007688.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | AK004659.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | L29468.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | AK162912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | BC007138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | NM_007688.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302149 | AK004659.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952895 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52103 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126040 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100159 | L29468.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100159 | BC007138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100159 | AK147174.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100159 | NM_007688.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100159 | AK004659.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | L29468.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | AK162912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | BC007138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | NM_007688.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | AK004659.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | L29468.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | AK162912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | BC007138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | NM_007688.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302149 | AK004659.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | L29468.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | AK162912.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | BC007138.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | NM_007688.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | AK004659.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | L29468.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | AK162912.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | BC007138.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | NM_007688.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302149 | AK004659.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28449503 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126040 | - | - |
| Whole Mouse Genome G4122A | A_51_P302149 | L29468.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | AK162912.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | BC007138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | NM_007688.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | AK004659.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | L29468.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | AK162912.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | BC007138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | NM_007688.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302149 | AK004659.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100159 | L29468.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100159 | BC007138.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100159 | AK147174.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100159 | NM_007688.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100159 | AK004659.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126040 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10169360 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10169361 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449511 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449509 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28449508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28449515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10011797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10276443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10011789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09856461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10276433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10245572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10276440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28449483 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10400352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400354 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10400351 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418066_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418067_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431432_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10400350 | - | - |
| Mouse Genome 430 2.0 Array | 1418067_at | - | - |
| Mouse Genome 430 2.0 Array | 1418066_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431432_at | - | - |
| Mouse Genome 430 2.0 Array | 1418067_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431432_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418066_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12632_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12632_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200012056.mm.1 | NM_007688.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200012056.mm.1 | NM_007688.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200012056.mm.1 | TC1200001757.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418066_PM_at | - | - |
| Mouse Expression Set 430 | 1418066_at | - | - |
| Mouse Expression Set 430 | 1418067_PM_at | - | - |
| Mouse Expression Set 430 | 1418067_at | - | - |
| Mouse Expression Set 430 | 1431432_at | - | - |
| Mouse Expression Set 430 | 1431432_PM_at | - | - |
| Murine Genome U74v2 Set | 97549_PM_at | - | - |
| Murine Genome U74v2 Set | 162088_PM_r_at | - | - |
| Murine Genome U74v2 Set | 97549_at | - | - |
| Murine Genome U74v2 Set | 162088_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418067_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418067_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418066_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418066_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418067_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418067_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418066_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418066_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431432_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431432_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200368 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200368 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 380239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200368 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249976 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241871 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249976 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241871 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002404.1_3-S | - | - |
| mouse-6 v1.1 (Illumina) | scl42461.3_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380239 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200368 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42461.3_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002404.1_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200368 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1241871 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1249976 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249976 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241871 | - | - |

Cfl2
Genomic Map