Assay Details
| Gene Symbol: Txnl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178604.3 | NP_848719.2 | 1 - 2 | 221 | 101 | |
| GenBank mRNA | AK011717.1 | - | 1 - 2 | 173 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011717.1, NM_178604.3 transcripts.
Set Membership: |
|

Txnl4a
Genomic Map