Assay Details
| Gene Symbol: Mboat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083341.1 | NP_001076810.1 | 2 - 3 | 319 | 86 | |
| NM_026037.3 | NP_080313.2 | 2 - 3 | 319 | 86 | ||
| GenBank mRNA | AB297383.1 | - | 2 - 3 | 218 | 86 | |
| AB305046.1 | - | 2 - 3 | 218 | 86 | ||
| AK012931.1 | - | 2 - 3 | 303 | 86 | ||
| AK031824.1 | - | 2 - 3 | 304 | 86 | ||
| AK076045.1 | - | 2 - 3 | 309 | 86 | ||
| BC025020.1 | - | 2 - 3 | 269 | 5364068 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031824.1 transcripts.
Set Membership: |
|

Mboat2
Genomic Map