| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800006002.mm.1 |
NM_013666.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006002.mm.1 |
NM_013666.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006002.mm.1 |
NM_153124.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006002.mm.1 |
NM_153124.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006002.mm.1 |
TC1800000827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005999.mm.1 |
NM_013666.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005999.mm.1 |
NM_013666.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005999.mm.1 |
NM_153124.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005999.mm.1 |
NM_153124.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005999.mm.1 |
TC1800000827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800006000.mm.1 |
NM_013666.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006000.mm.1 |
NM_013666.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006000.mm.1 |
NM_153124.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006000.mm.1 |
NM_153124.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800006000.mm.1 |
TC1800000827.mm.1 |
MTA TC |