Assay Details
| Gene Symbol: Smad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008540.2 | NP_032566.2 | 9 - 10 | 1578 | 63 | |
| GenBank mRNA | AB221585.1 | - | 8 - 9 | 1138 | 63 | |
| AK004804.1 | - | 8 - 9 | 942 | 63 | ||
| AY493561.1 | - | 8 - 9 | 1138 | 63 | ||
| BC046584.1 | - | 9 - 10 | 1578 | 6313280 | 63 | |
| GU072917.1 | - | 6 - 7 | 901 | 63 | ||
| GU072918.1 | - | 5 - 6 | 850 | 63 | ||
| GU072920.1 | - | 8 - 9 | 1138 | 63 | ||
| U79748.1 | - | 8 - 9 | 1458 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smad4
Genomic Map