Assay Details
| Gene Symbol: Stard6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289648.1 | NP_001276577.1 | 3 - 4 | 273 | 91 | |
| NM_001289649.1 | NP_001276578.1 | 3 - 4 | 252 | 91 | ||
| NM_001289650.1 | NP_001276579.1 | 3 - 4 | 287 | 91 | ||
| NM_029019.4 | NP_083295.1 | 3 - 4 | 342 | 91 | ||
| GenBank mRNA | AK014759.1 | - | 3 - 4 | 253 | 91 | |
| AK016984.1 | - | 3 - 4 | 253 | 91 | ||
| BC087893.1 | - | 3 - 4 | 264 | 6774832 | 91 | |
| JF440953.1 | - | 1 - 2 | 61 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014759.1, AK016984.1, BC087893.1, JF440953.1, NM_001289648.1, NM_001289649.1, NM_001289650.1, NM_029019.4 transcripts.
Set Membership: |
|

Stard6
Genomic Map