Assay Details
| Gene Symbol: Spink6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013797.1 | NP_001013819.1 | 4 - 5 | 343 | 84 | |
| GenBank mRNA | AK028032.1 | - | 4 - 5 | 343 | 84 | |
| BC147471.1 | - | 3 - 4 | 239 | 9056113 | 84 | |
| BC171990.1 | - | 3 - 4 | 239 | 9054276 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432607 | NM_001013797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432607 | AK028032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432607 | NM_001013797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432607 | AK028032.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825260 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96562 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639720 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633309 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65109 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131715 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137091 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432607 | NM_001013797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432607 | AK028032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432607 | NM_001013797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432607 | AK028032.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31665010 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08983482 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137091 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131715 | - | - |
| Whole Mouse Genome G4122A | A_51_P432607 | NM_001013797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432607 | AK028032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432607 | NM_001013797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432607 | AK028032.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126296 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131715 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31665009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13969834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13906569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13797777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13983901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13983902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31665022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10455442 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455444 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10455441 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800003315.mm.1 | NM_001013797.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003315.mm.1 | NM_001013797.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003315.mm.1 | TC1800000450.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800003314.mm.1 | NM_001013797.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003314.mm.1 | NM_001013797.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003314.mm.1 | TC1800000450.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 760040 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 103120114 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2577593 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103120114 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103120114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2996988 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2996988 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2996983 | - | - |

Spink6
Genomic Map