Assay Details
| Gene Symbol: 9130209A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033453.1 | 2 - 3 | 216 | 83 | ||
| GenBank mRNA | AK020276.1 | - | 2 - 3 | 217 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A |
Sema6a |
9330158E07, A730020P05Rik, AI851735, Sema6A-1, Semaq, VIa, b2b997Clo |
-82.6kb | Anti-sense | Chr. 18 - 47245254 - 47371487 on NCBI Build GRCm38 |
| Downstream | predicted gene 5095 | Gm5095 | A730092B10 | 81.2kb | Sense | Chr. 18 - 47537735 - 47718636 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525793 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394909 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65303 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03468617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13643666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13754580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14052847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13806433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13626022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13637426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13626020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14052846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13772662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13904566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13626025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13806434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13626030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14025270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31682729 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1800003572.mm.1 | NR_033453.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003572.mm.1 | NR_033453.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003572.mm.1 | TC1800000494.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800003575.mm.1 | NR_033453.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003575.mm.1 | NR_033453.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003575.mm.1 | TC1800000494.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800003574.mm.1 | NR_033453.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003574.mm.1 | NR_033453.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003574.mm.1 | TC1800000494.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800003573.mm.1 | NR_033453.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003573.mm.1 | NR_033453.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003573.mm.1 | TC1800000494.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 338991 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101230731 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2736838 | - | - |

9130209A04Rik
Genomic Map