Assay Details
| Gene Symbol: Tcof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198984.1 | NP_001185913.1 | 14 - 15 | 2250 | 75 | |
| NM_011552.3 | NP_035682.1 | 14 - 15 | 2250 | 75 | ||
| GenBank mRNA | AF001794.1 | - | 14 - 15 | 2194 | 75 | |
| AK089518.1 | - | 14 - 15 | 2218 | 75 | ||
| AK170990.1 | - | 2 - 3 | 146 | 75 | ||
| AK171978.1 | - | 2 - 3 | 146 | 75 | ||
| BC021514.1 | - | 14 - 15 | 2255 | 5351783 | 75 | |
| BC046608.1 | - | 13 - 14 | 2060 | 75 | ||
| BC057342.1 | - | 14 - 15 | 2203 | 6825735 | 75 | |
| BC060105.1 | - | 14 - 15 | 2203 | 75 | ||
| U81030.1 | - | 14 - 15 | 2138 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcof1
Genomic Map