Assay Details
| Gene Symbol: Tcof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198984.1 | NP_001185913.1 | 22 - 23 | 3526 | 135 | |
| NM_011552.3 | NP_035682.1 | 21 - 22 | 3418 | 135 | ||
| GenBank mRNA | AF001794.1 | - | 21 - 22 | 3362 | 135 | |
| AK089518.1 | - | 22 - 23 | 137 | |||
| BC021514.1 | - | 22 - 23 | 3541 | 5351783 | 135 | |
| BC046608.1 | - | 20 - 21 | 3228 | 135 | ||
| BC052669.1 | - | 22 - 23 | 3498 | 135 | ||
| BC057342.1 | - | 22 - 23 | 3478 | 6825735 | 135 | |
| BC060105.1 | - | 21 - 22 | 3371 | 135 | ||
| U81030.1 | - | 21 - 22 | 3306 | 135 | ||
More Information
Set Membership: |
|

Tcof1
Genomic Map