Assay Details
| Gene Symbol: Tcof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198984.1 | NP_001185913.1 | 10 - 11 | 1610 | 162 | |
| NM_011552.3 | NP_035682.1 | 10 - 11 | 1610 | 162 | ||
| GenBank mRNA | AF001794.1 | - | 10 - 11 | 1554 | 162 | |
| AK089518.1 | - | 10 - 11 | 1577 | 162 | ||
| BC021514.1 | - | 10 - 11 | 1615 | 5351783 | 162 | |
| BC052669.1 | - | 10 - 11 | 1582 | 162 | ||
| BC057342.1 | - | 10 - 11 | 1563 | 6825735 | 162 | |
| BC060105.1 | - | 10 - 11 | 1563 | 162 | ||
| U81030.1 | - | 10 - 11 | 1498 | 162 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcof1
Genomic Map