Assay Details
| Gene Symbol: Tcof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198984.1 | NP_001185913.1 | 25 - 26 | 4195 | 87 | |
| NM_011552.3 | NP_035682.1 | 24 - 25 | 4087 | 87 | ||
| GenBank mRNA | AK171978.1 | - | 13 - 14 | 2095 | 88 | |
| BC057342.1 | - | 25 - 26 | 4147 | 6825735 | 87 | |
| U81030.1 | - | 24 - 25 | 3975 | 87 | ||
More Information
Set Membership: |
|

Tcof1
Genomic Map