Assay Details
| Gene Symbol: Cdc42se1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038708.3 | NP_001033797.1 | 3 - 4 | 486 | 92 | |
| NM_172395.3 | NP_765983.1 | 3 - 4 | 856 | 92 | ||
| GenBank mRNA | AK004859.1 | - | 3 - 4 | 501 | 92 | |
| AK008084.1 | - | 2 - 3 | 527 | 92 | ||
| AK019052.1 | - | 3 - 4 | 498 | 92 | ||
| AK028573.1 | - | 3 - 4 | 498 | 92 | ||
| AK213146.1 | - | 1 - 2 | 72 | 92 | ||
| BC029194.1 | - | 3 - 4 | 474 | 93 | ||
| BC060964.1 | - | 3 - 4 | 478 | 6743495 | 92 | |
| CF105339.1 | - | 3 - 4 | 486 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cdc42se1
Genomic Map