Assay Details
| Gene Symbol: Mthfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010840.3 | NP_034970.2 | 1 - 2 | 57 | 56 | |
| GenBank mRNA | AF404271.1 | - | 1 - 2 | 57 | 56 | |
| AK030192.1 | - | 1 - 2 | 52 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF404271.1, AK030192.1, NM_010840.3 transcripts.
Set Membership: |
|

Mthfr
Genomic Map