Assay Details
| Gene Symbol: Masp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003893.2 | NP_001003893.1 | 9 - 10 | 1265 | 74 | |
| GenBank mRNA | AB009459.1 | - | 9 - 10 | 1255 | 74 | |
| AK050052.1 | - | 9 - 10 | 1242 | 74 | ||
| AK149421.1 | - | 8 - 9 | 1319 | 74 | ||
| BC013893.1 | - | 8 - 9 | 1239 | 4211293 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Masp2
Genomic Map