Assay Details
| Gene Symbol: Masp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003893.2 | NP_001003893.1 | 10 - 11 | 1335 | 102 | |
| GenBank mRNA | AB009459.1 | - | 10 - 11 | 1325 | 102 | |
| AK050052.1 | - | 10 - 11 | 1312 | 102 | ||
| AK149421.1 | - | 9 - 10 | 1389 | 102 | ||
| BC013893.1 | - | 9 - 10 | 1309 | 4211293 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Masp2
Genomic Map