Assay Details
| Gene Symbol: Gnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160016.1 | NP_001153488.1 | 2 - 3 | 275 | 71 | |
| NM_008142.4 | NP_032168.1 | 2 - 3 | 275 | 71 | ||
| GenBank mRNA | AK163844.1 | - | 2 - 3 | 264 | 71 | |
| BC013058.1 | - | 2 - 3 | 266 | 3964965 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK163844.1, BC013058.1, NM_001160016.1, NM_008142.4 transcripts.
Set Membership: |
|

Gnb1
Genomic Map