Assay Details
| Gene Symbol: Dffb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007859.4 | NP_031885.3 | 3 - 4 | 532 | 113 | |
| GenBank mRNA | AB009377.1 | - | 3 - 4 | 440 | 113 | |
| AK077619.1 | - | 3 - 4 | 578 | 113 | ||
| AK089267.1 | - | 3 - 4 | 509 | 113 | ||
| BC053052.1 | - | 3 - 4 | 462 | 6403143 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dffb
Genomic Map