Assay Details
| Gene Symbol: Slc38a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037717.3 | NP_001032806.2 | 13 - 14 | 1135 | 80 | |
| GenBank mRNA | BC043932.1 | - | 6 - 7 | 462 | 80 | |
| BC157917.1 | - | 13 - 14 | 1084 | 9007345 | 80 | |
| BC157928.1 | - | 13 - 14 | 1084 | 9088101 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a6
Genomic Map