Assay Details
| Gene Symbol: Slc38a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037717.3 | NP_001032806.2 | 1 - 2 | 186 | 115 | |
| GenBank mRNA | AK054341.1 | - | 1 - 2 | 140 | 115 | |
| AK140045.1 | - | 1 - 2 | 179 | 115 | ||
| BC157917.1 | - | 1 - 2 | 135 | 9007345 | 115 | |
| BC157928.1 | - | 1 - 2 | 135 | 9088101 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a6
Genomic Map