Assay Details
| Gene Symbol: Slc38a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037717.3 | NP_001032806.2 | 4 - 5 | 446 | 117 | |
| GenBank mRNA | BC157917.1 | - | 4 - 5 | 395 | 9007345 | 117 |
| BC157928.1 | - | 4 - 5 | 395 | 9088101 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a6
Genomic Map